Anti-GFP Rabbit Polyclonal Antibody

Supplier: Abnova
PAB0863
10676-314EA 329 USD
10676-314
Anti-GFP Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against GFP from Aequorea victoria.

Type: Primary
Antigen: GFP
Clonality: Unconjugated
Clone: Polyclonal
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR