Anti-SPA17 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB0869
10681-222EA 329 USD
10681-222
Anti-SPA17 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against a synthetic peptide of SPA17.

Recommended Dilutions: Western Blot: 1:200

Type: Primary
Antigen: SPA17
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR