Anti-Glu-Glu tag Polyclonal Antibody

Supplier: Abnova
PAB0881
10681-892EA 329 USD
10681-892
Anti-Glu-Glu tag Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against Glu-Glu tag.

Recommended Dilutions: Western Blot: 1:400

Type: Primary
Antigen: Glu-Glu tag
Clonality: Unconjugated
Clone: Polyclonal
Conjugation:
Epitope:
Host:
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR