Anti-VSV Polyclonal Antibody

Supplier: Abnova
PAB0886
10677-462EA 406.15 USD
10677-462
Anti-VSV Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against VSV-G tag.

Recommended Dilutions: Western Blot: 1:400

Type: Primary
Antigen: VSV
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host:
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR