Anti-MBP Polyclonal Antibody

Supplier: Abnova
PAB0893
10676-838EA 406.15 USD
10676-838
Anti-MBP Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against MBP (Maltose Binding Protein).

Recommended Dilutions: Western Blot: 1:400

Type: Primary
Antigen: MBP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host:
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR