Anti-Biotin Polyclonal Antibody

Supplier: Abnova
PAB0895
10673-262EA 329 USD
10673-262
Anti-Biotin Polyclonal Antibody
Antibodies
Goat polyclonal antibody raised against a Biotinylated to carrier protein.

Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: Biotin
Clonality: Unconjugated
Clone: Polyclonal
Conjugation:
Epitope:
Host:
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR