Anti-BSA Sheep Polyclonal Antibody

Supplier: Abnova
PAB0903
10673-406EA 426.15 USD
10673-406
Anti-BSA Sheep Polyclonal Antibody
Antibodies
Sheep polyclonal antibody raised against Bovine Serum Albumin, BSA.

Type: Primary
Antigen: BSA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype:
Reactivity: 0
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR