Anti-FBXO43 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB21674
10714-508EA 428.55 USD
10714-508
Anti-FBXO43 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against recombinant FBXO43.

Recommended Dilutions: Immunohistochemistry: 1:200-1:500; Western Blot: 1:250-1:500

Type: Primary
Antigen: FBXO43
Clonality: Unconjugated
Clone: Polyclonal
Conjugation:
Epitope:
Host: IgG
Isotype: Rabbit
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR