Anti-SLC35B2 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB22320
10737-656EA 428.55 USD
10737-656
Anti-SLC35B2 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against recombinant SLC35B2.

Recommended Dilutions: Immunohistochemistry: 1:10-1:20

Type: Primary
Antigen: SLC35B2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR