Anti-ECT2 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB10006
10674-520EA 786.15 USD
10674-520
Anti-ECT2 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic phosphopeptide of ECT2.

Recommended Dilutions: ELISA: 1:10000-1:40000; Western Blot: 1:1000-1:4000

Type: Primary
Antigen: ECT2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Chicken, Chimpanzee, Dog, Human, Mouse, Rat, Zebrafish
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR