Anti-EGFR Rabbit Polyclonal Antibody

Supplier: Abnova
PAB10009
10674-546EA 786.15 USD
10674-546
Anti-EGFR Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic phosphopeptide of EGFR.

Recommended Dilutions: ELISA: 1:4000-1:20000; Western Blot: 1:250-1:1500; Immunohistochemistry: 5 ?g/ml

Type: Primary
Antigen: EGFR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR