Anti-FBXO9 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB10012
10714-528EA 638 USD
10714-528
Anti-FBXO9 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of FBXO9.

Recommended Dilutions: ELISA: 1:5000-1:25000; Western Blot: 1:500-1:2000

Type: Primary
Antigen: FBXO9
Clonality: Unconjugated
Clone: Polyclonal
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Bovine, Chicken, Chimpanzee, Clawed frog, Dog, Human, Mouse, Rat, Zebrafish
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR