Anti-FKBP8 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB10013
10714-824EA 638 USD
10714-824
Anti-FKBP8 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of FKBP8.

Recommended Dilutions: ELISA: 1:25000; Western Blot: 1:500-1:2000

Type: Primary
Antigen: FKBP8
Clonality: Unconjugated
Clone: Polyclonal
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR