Anti-APC2 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB10021
10689-558EA 638 USD
10689-558
Anti-APC2 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of APC2.

Recommended Dilutions: ELISA: 1:2000-1:10000; Western Blot: 1:500-1:1000

Type: Primary
Antigen: APC2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR