Anti-AURKB Rabbit Polyclonal Antibody

Supplier: Abnova
PAB10025
10692-648EA 638 USD
10692-648
Anti-AURKB Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of AURKB.

Recommended Dilutions: ELISA: 1:5000-1:20000; Western Blot: 1:500-1:2000

Type: Primary
Antigen: AURKB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bovine, Chimpanzee, Dog, Human, Mouse, Pig, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR