Anti-FANCA Rabbit Polyclonal Antibody

Supplier: Abnova
PAB10045
10714-280EA 786.15 USD
10714-280
Anti-FANCA Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of FANCA.

Recommended Dilutions: ELISA: 1:15000-1:60000; Western Blot: 1:500-1:3000

Type: Primary
Antigen: FANCA
Clonality: Unconjugated
Clone: Polyclonal
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Chimpanzee, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR