Anti-L-Dihydroxyphenylalanine (L-DOPA) Polyclonal Antibody

Supplier: Abnova
PAB0031
10723-474EA 725 USD
10723-474
Anti-L-Dihydroxyphenylalanine (L-DOPA) Polyclonal Antibody
Rabbit polyclonal antibody raised against L-DOPA.

Recommended Dilutions: ELISA: 1:2000-1:10000; Immunohistochemistry: 1:1000-1:5000

Type: Primary
Antigen: L-Dihydroxyphenylalanine (L-DOPA)
Clonality: Unconjugated
Clone: Polyclonal
Conjugation:
Epitope:
Host:
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR