Anti-NEXN Rabbit Polyclonal Antibody

Supplier: Abnova
PAB20543
10727-004EA 527.69 USD
10727-004
Anti-NEXN Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against recombinant NEXN.

Recommended Dilutions: Immunohistochemistry: 1:500-1:1000

Type: Primary
Antigen: NEXN
Clonality: Unconjugated
Clone: Polyclonal
Conjugation:
Epitope:
Host: IgG
Isotype: Rabbit
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR