Anti-GSK3B Rabbit Polyclonal Antibody

Supplier: Abnova
PAB10055
10674-692EA 638 USD
10674-692
Anti-GSK3B Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic phosphopeptide of GSK3B.

Recommended Dilutions: ELISA: 1:10000-1:50000; Western Blot: 1:500-1:3000; Immunohistochemistry: 1:200-1:1000; Immunofluorescence: 1:200-1:1000

Type: Primary
Antigen: GSK3B
Clonality: Unconjugated
Clone: Polyclonal
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Chicken, Dog, Frog, Human, Rat, Zebrafish
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR