Anti-RAD23A Goat Polyclonal Antibody

Supplier: Abnova
PAB10057
10734-088EA 786.15 USD
10734-088
Anti-RAD23A Goat Polyclonal Antibody
Antibodies
Goat polyclonal antibody raised against synthetic peptide of RAD23A.

Recommended Dilutions: ELISA: 1:4000-1:16000; Western Blot: 1:500-1:2000

Type: Primary
Antigen: RAD23A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR