Anti-Caproic acid Polyclonal Antibody

PAB0041
10704-898EA 1036.92 USD
10704-898
Anti-Caproic acid Polyclonal Antibody
Rat polyclonal antibody raised against Caproic acid.

Recommended Dilutions: ELISA: 1:2000-1:10000; Immunohistochemistry: 1:1000-1:5000

Type: Primary
Antigen: Caproic acid
Clonality: Unconjugated
Clone: Polyclonal
Conjugation:
Epitope:
Host:
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR