Anti-SMARCAL1 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB10063
10738-728EA 638 USD
10738-728
Anti-SMARCAL1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of SMARCAL1.

Recommended Dilutions: ELISA: 1:16000-1:64000; Western Blot: 1:500-1:3000

Type: Primary
Antigen: SMARCAL1
Clonality: Unconjugated
Clone: Polyclonal
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR