Anti-ST8SIA6 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB20556
10739-576EA 428.55 USD
10739-576
Anti-ST8SIA6 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against recombinant ST8SIA6.

  • Recommended Dilutions: Immunohistochemistry: 1:50-1:200

Type: Primary
Antigen: ST8SIA6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR