Anti-5-Methoxytryptamine Polyclonal Antibody

PAB0050
10692-242EA 910.77 USD
10692-242
Anti-5-Methoxytryptamine Polyclonal Antibody
Rabbit polyclonal antibody raised against 5-methoxytryptamine.

Recommended Dilutions: ELISA: 1:2000-1:10000; Immunohistochemistry: 1:1000-1:5000

Type: Primary
Antigen: 5-Methoxytryptamine
Clonality: Unconjugated
Clone: Polyclonal
Conjugation:
Epitope:
Host:
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR