Anti-PARK7 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB10070
10733-366EA 638 USD
10733-366
Anti-PARK7 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of PARK7.

Recommended Dilutions: ELISA: 1:10000-1:50000; Western Blot: 1:500-1:2000; Immunohistochemistry: 2-5 ?g/ml

Type: Primary
Antigen: PARK7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Chicken, Dog, Frog, Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR