Anti-5,7-Dihydroxytryptamine Polyclonal Antibody

PAB0054
10687-458EA 1036.92 USD
10687-458
Anti-5,7-Dihydroxytryptamine Polyclonal Antibody
Rabbit polyclonal antibody raised against 5,7-dihydroxytryptamine.

Recommended Dilutions: ELISA: 1:2000-1:10000; Immunohistochemistry: 1:1000-1:5000

Type: Primary
Antigen: 5,7-Dihydroxytryptamine
Clonality: Unconjugated
Clone: Polyclonal
Conjugation:
Epitope:
Host: IgG
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR