Anti-NEK2 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB10072
10726-916EA 786.15 USD
10726-916
Anti-NEK2 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of NEK2.

Recommended Dilutions: ELISA: 1:2000-1:8000

Type: Primary
Antigen: NEK2
Clonality: Unconjugated
Clone: Polyclonal
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR