Anti-RRP1B Rabbit Polyclonal Antibody

Supplier: Abnova
PAB21152
10732-040EA 428.55 USD
10732-040
Anti-RRP1B Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against recombinant RRP1B.

Recommended Dilutions: Immunohistochemistry: 1:500-1:1000; Immunofluorescence: 1-4 ?g/ml

Type: Primary
Antigen: RRP1B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR