Anti-NO_2-Serotonin Polyclonal Antibody

PAB0060
10727-320EA 910.77 USD
10727-320
Anti-NO_2-Serotonin Polyclonal Antibody
Rat polyclonal antibody raised against NO2-Serotonin.

Recommended Dilutions: ELISA: 1:2000-1:10000; Immunohistochemistry: 1:1000-1:5000

Type: Primary
Antigen: NO_2-Serotonin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host:
Isotype:
Reactivity: 0
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR