Anti-XIRP1 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB21166
10744-818EA 428.55 USD
10744-818
Anti-XIRP1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against recombinant XIRP1.

Recommended Dilutions: Immunohistochemistry: 1:50-1:200; Immunofluorescence: 1-4 ?g/ml

Type: Primary
Antigen: XIRP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR