Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-SLC38A6 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB21170
10737-690
Anti-SLC38A6 Rabbit Polyclonal Antibody
Rabbit polyclonal antibody raised against recombinant SLC38A6.

Recommended Dilutions: Immunohistochemistry: 1:50-1:200

Type: Primary
Antigen: SLC38A6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now


 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty