Anti-CERS2 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB10110
10685-148EA 428 USD
10685-148
Anti-CERS2 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of LASS2.

Recommended Dilutions: Western Blot: 1:400

Type: Primary
Antigen: CERS2
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR