Anti-LY86 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB10120
10724-250EA 746.62 USD
10724-250
Anti-LY86 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of LY86.

Recommended Dilutions: Western Blot: 0.5 to 1 ?g/ml

Type: Primary
Antigen: LY86
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR