Anti-TWISTNB Rabbit Polyclonal Antibody

Supplier: Abnova
PAB21189
10743-390EA 428.55 USD
10743-390
Anti-TWISTNB Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against recombinant TWISTNB.

Recommended Dilutions: Immunohistochemistry: 1:200-1:500; Immunofluorescence: 1-4 ?g/ml

Type: Primary
Antigen: TWISTNB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR