Anti-GPR68 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB10144
10716-272EA 849.43 USD
10716-272
Anti-GPR68 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against a recombinant GPR68.

Recommended Dilutions: Western Blot: 5 to 10 ?g/ml

Type: Primary
Antigen: GPR68
Clonality: Unconjugated
Clone: Polyclonal
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR