Anti-TP73 Polyclonal Antibody

Supplier: Abnova
PAB10146
10742-516EA 428 USD
10742-516
Anti-TP73 Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against partial recombinant TP73.

Recommended Dilutions: Western Blot: 5 to 10 ?g/ml

Type: Primary
Antigen: TP73
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host:
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR