Anti-EIF2AK3 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB10175
10711-770EA 638 USD
10711-770
Anti-EIF2AK3 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against partial recombinant Eif2ak3.

Recommended Dilutions: Western Blot: 1:500-1:3000; Immunoprecipitation: 10-30?l)

Type: Primary
Antigen: EIF2AK3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR