Anti-CAT Rabbit Polyclonal Antibody

Supplier: Abnova
PAB10185
10705-308EA 527.69 USD
10705-308
Anti-CAT Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of Chloramphenicol acetyl transferase.

Recommended Dilutions: ELISA: 1:5000-1:25000

Type: Primary
Antigen: CAT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR