Anti-Collagen Type V Rabbit Polyclonal Antibody

Supplier: Abnova
PAB10194
10708-736EA 746.62 USD
10708-736
Anti-Collagen Type V Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against native Collagen Type V.

Recommended Dilutions: ELISA: 1:5000-1:10000; Western Blot: 1:5000-1:10000; Immunohistochemistry: 1:50-1:200

Type: Primary
Antigen: Collagen Type V
Clonality: Unconjugated
Clone: Polyclonal
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Bovine, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR