Anti-IL21R Rabbit Polyclonal Antibody

Supplier: Abnova
PAB0364
10719-152EA 428.44 USD
10719-152
Anti-IL21R Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of IL21R.

Type: Primary
Antigen: IL21R
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR