Anti-MKI67IP Rabbit Polyclonal Antibody

Supplier: Abnova
PAB0369
10676-850EA 428.44 USD
10676-850
Anti-MKI67IP Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of MKI67IP.

Type: Primary
Antigen: MKI67IP
Clonality: Unconjugated
Clone: Polyclonal
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR