Anti-B3GAT2 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB20580
10690-656EA 428.55 USD
10690-656
Anti-B3GAT2 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against recombinant B3GAT2.

Recommended Dilutions: Immunohistochemistry: 1:20-1:50

Type: Primary
Antigen: B3GAT2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR