Anti-USP36 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB20583
10743-912EA 428.55 USD
10743-912
Anti-USP36 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against recombinant USP36.

Recommended Dilutions: Immunohistochemistry: 1:20-1:50; Western Blot: 1:250-1:500; Immunofluorescence: 1-4 ?g/ml

Type: Primary
Antigen: USP36
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR