Anti-SCGB2A2 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB0129
10732-386EA 638.32 USD
10732-386
Anti-SCGB2A2 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide conjugated to KLH.

Recommended Dilutions: Western Blot: 1 ?g/ml; Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 1 ?g/ml

Type: Primary
Antigen: SCGB2A2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR