Anti-FCGRT Rabbit Polyclonal Antibody

Supplier: Abnova
PAB20589
10720-216EA 428.55 USD
10720-216
Anti-FCGRT Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against recombinant FCGRT.

Recommended Dilutions: Immunohistochemistry: 1:50-1:200; Western Blot: 1:250-1:500

Type: Primary
Antigen: FCGRT
Clonality: Unconjugated
Clone: Polyclonal
Conjugation:
Epitope:
Host: IgG
Isotype: Rabbit
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR