Anti-DIAPH2 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB10201
10710-692EA 638 USD
10710-692
Anti-DIAPH2 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of DIAPH2.

Recommended Dilutions: ELISA: 1:12000-1:60000; Western Blot: 1:1000-1:6000

Type: Primary
Antigen: DIAPH2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR