Anti-EIF3E Rabbit Polyclonal Antibody

Supplier: Abnova
PAB10203
10711-790EA 638 USD
10711-790
Anti-EIF3E Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of Eif3e.

Recommended Dilutions: ELISA: 1:90000; Western Blot: 1:1000

Type: Primary
Antigen: EIF3E
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bovine, Dog, Human, Monkey, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR