Anti-NCAM2 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB0134
10726-702EA 747.11 USD
10726-702
Anti-NCAM2 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against mouse NCAM-2 (442 a.a. ~ 685 a.a) partial recombinant protein.

Recommended Dilutions: Western Blot: 1:1000; Immunohistochemistry: 1:500

Type: Primary
Antigen: NCAM2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR