Anti-FBX4 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB10205
10714-484EA 638 USD
10714-484
Anti-FBX4 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of FBX4.

Recommended Dilutions: Western Blot: 1:500-1:3000

Type: Primary
Antigen: FBX4
Clonality: Unconjugated
Clone: Polyclonal
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR