Anti-KLH Polyclonal Antibody

Supplier: Abnova
PAB0144
10685-122EA 527.69 USD
10685-122
Anti-KLH Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against purified KLH.

Recommended Dilutions: Western Blot: 1:10000; ELISA: 1:10000

Type: Primary
Antigen: KLH
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host:
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR